Thermo Scientific Chemicals

2,6-Dichloropyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27406.MD
250 mg, Each
Thermo Scientific Chemicals

2,6-Dichloropyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27406.MD
250 mg, Each
Quantity
Catalog number: H27406.MD
also known as H27406-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
148493-34-9
IUPAC Name
(2,6-dichloropyridin-3-yl)boronic acid
Molecular Formula
C5H4BCl2NO2
InChI Key
XBBLBQZAVMHEER-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(Cl)N=C1Cl
Form
Powder
Melting Point (clear melt)
91.0-103.0?C
Assay (HPLC)
≥94.0%
Proton NMR
Conforms to structure
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text