Thermo Scientific Chemicals

1-Methyl-4-(4-piperidinyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: H27408.03
1 g, Each
Thermo Scientific Chemicals

1-Methyl-4-(4-piperidinyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: H27408.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H27408.03
also known as H27408-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53617-36-0
IUPAC Name
1-methyl-4-(piperidin-4-yl)piperazine
Molecular Formula
C10H21N3
InChI Key
MRYYJGQKVGZGSB-UHFFFAOYSA-N
SMILES
CN1CCN(CC1)C1CCNCC1
Form
Powder, lumpy or fused solid
Melting Point (clear melt)
52-57?C
Appearance (Color)
White to cream to orange
Assay (GC)
≥97.5%

Description

It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.

Solubility
Soluble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids. Store under dry inert gas. Refrigerated (2-4°C).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text