It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.
Solubility
Soluble in water.
Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids. Store under dry inert gas. Refrigerated (2-4°C).
RUO – Research Use Only
Xiaoqiang Qiao,; Liangliang Sun,; Lingfan Chen,; Yuan Zhou,; Kaiguang Yang,; Zhen Liang,; Lihua Zhann,; Yukui Zhang. Piperazines for peptide carboxyl group derivatization: effect of derivatization reagents and properties of peptides on signal enhancement in matrix-assisted laser desorption/ionization mass spectrometry. Rapid Communications in Mass Spectrometry. 2011, 25 (5),639-646.