Thermo Scientific Chemicals

4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%, Thermo Scientific Chemicals

Catalog number: H27420.ME
500 mg, Each
Thermo Scientific Chemicals

4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%, Thermo Scientific Chemicals

Catalog number: H27420.ME
500 mg, Each
Quantity
Catalog number: H27420.ME
also known as H27420-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
856405-77-1
IUPAC Name
di-tert-butyl[5-(di-tert-butylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]phosphane
Molecular Formula
C31H48OP2
InChI Key
ZEIZANJFJXHMNS-UHFFFAOYSA-N
SMILES
CC(C)(C)P(C1=CC=CC2=C1OC1=C(C=CC=C1P(C(C)(C)C)C(C)(C)C)C2(C)C)C(C)(C)C
Appearance (Color)
White
Assay by NMR
>98.5% by 31P NMR
Proton NMR
Conforms to structure
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text