trans-4-Hydroxy-D-proline, 97%, Thermo Scientific Chemicals
trans-4-Hydroxy-D-proline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Hydroxy-D-proline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H27421.03
also known as H27421-03
Price (USD)/ Each
245.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
245.00
Add to cart
trans-4-Hydroxy-D-proline, 97%, Thermo Scientific Chemicals
Catalog numberH27421.03
Price (USD)/ Each
245.00
-
Add to cart
Chemical Identifiers
CAS3398-22-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder, crystalline powder, crystals and/or chunks
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
Optical Rotation+68? to +82? (c=1, water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only