(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- Joan Albert,; Jaume Granell,; Joaquim Sales,; Merce Font-Bardia,; Xavier Solans. Optically Active Exocyclic Cyclopalladated Derivatives of Benzylidene-(R)-(1-phenylethyl)amines: Syntheses and X-ray Molecular Structures of [Pd(2-{(E)-(R)-CHMeN:CH-2',6'-Cl2C6H3}C6H4)Cl(PPh3)] and [Pd(2-{(Z)-(R)-CHMeN:CH-2',6'-F2C6H3}C6H4)I(PPh3)]. Organometallics. 1995, 14 (3), 1393-1404.