Thermo Scientific Chemicals

Tricyclohexylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H27428.14
25 g, Each
Thermo Scientific Chemicals

Tricyclohexylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H27428.14
25 g, Each
Quantity
Catalog number: H27428.14
also known as H27428-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58656-04-5
IUPAC Name
tetrafluoroboranuide; tricyclohexylphosphanium
Molecular Formula
C18H34BF4P
InChI Key
MYSMMEUXKHJYKH-UHFFFAOYSA-O
SMILES
F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1
Appearance (Color)
White
Form
Crystalline powder or powder
Assay (Aqueous acid-base Titration)
>98.5 to <101.5%
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure

Description

Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text