N-Acetyl-DL-leucine, 99%, Thermo Scientific Chemicals
N-Acetyl-DL-leucine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetyl-DL-leucine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number H27441.22
also known as H27441-22
Price (USD)/ Each
541.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
541.00
Add to cart
N-Acetyl-DL-leucine, 99%, Thermo Scientific Chemicals
Catalog numberH27441.22
Price (USD)/ Each
541.00
-
Add to cart
Chemical Identifiers
CAS99-15-0
IUPAC Name2-acetamido-4-methylpentanoic acid
Molecular FormulaC8H15NO3
InChI KeyWXNXCEHXYPACJF-UHFFFAOYNA-N
SMILESCC(C)CC(NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)156.0-162.0?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only