Thermo Scientific Chemicals

4-Iodobiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: H27453.06
5 g, Each
Thermo Scientific Chemicals

4-Iodobiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: H27453.06
5 g, Each
Quantity
Catalog number: H27453.06
also known as H27453-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1591-31-7
IUPAC Name
4-iodo-1,1'-biphenyl
Molecular Formula
C12H9I
InChI Key
NXYICUMSYKIABQ-UHFFFAOYSA-N
SMILES
IC1=CC=C(C=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
108.0-118.0?C

Description

4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text