Thermo Scientific Chemicals

6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27463.03
1 g, Each
Thermo Scientific Chemicals

6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27463.03
1 g, Each
Quantity
Catalog number: H27463.03
also known as H27463-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1073354-27-4
IUPAC Name
N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Formula
C18H23BN2O2
InChI Key
BWIHFPRFKBKWMU-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CN=C(NCC2=CC=CC=C2)C=C1
Appearance (Color)
Pale yellow to yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥94.0%
Melting Point (clear melt)
129.0-141.0?C

Description

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text