6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27463.MD
also known as H27463-MD
Price (USD)/ Each
163.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
163.00
Add to cart
6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog numberH27463.MD
Price (USD)/ Each
163.00
-
Add to cart
Chemical Identifiers
CAS1073354-27-4
IUPAC NameN-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular FormulaC18H23BN2O2
InChI KeyBWIHFPRFKBKWMU-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CN=C(NCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
Melting Point (clear melt)129.0-141.0?C
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only