4-(Aminomethyl)benzonitrile hydrochloride, 97%, Thermo Scientific Chemicals
4-(Aminomethyl)benzonitrile hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Aminomethyl)benzonitrile hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27473.06
also known as H27473-06
Price (USD)/ Each
82.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
82.10
Add to cart
4-(Aminomethyl)benzonitrile hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH27473.06
Price (USD)/ Each
82.10
-
Add to cart
Chemical Identifiers
CAS15996-76-6
IUPAC Namehydrogen 4-(aminomethyl)benzonitrile chloride
Molecular FormulaC8H9ClN2
InChI KeyQREZLLYPLRPULF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
Melting Point (clear melt)270-282?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only