D-Isoleucine, 98%
Thermo Scientific Chemicals

D-Isoleucine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27488.MD
also known as H27488-MD
Price (USD)/ Each
279.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
279.00
Add to cart
D-Isoleucine, 98%
Catalog numberH27488.MD
Price (USD)/ Each
279.00
-
Add to cart
Chemical Identifiers
CAS319-78-8
IUPAC Name(2R,3R)-2-amino-3-methylpentanoic acid
Molecular FormulaC6H13NO2
InChI KeyAGPKZVBTJJNPAG-LXDRQGDMNA-N
SMILESCC[C@@H](C)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Appearance (Color)White to cream
Optical Rotation-39.0? ? 2.0? (c=5, 5M aq. HCl)
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only