1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals
1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number H27548.18
also known as H27548-18
Price (USD)/ Each
422.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
422.00
Add to cart
1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals
Catalog numberH27548.18
Price (USD)/ Each
422.00
-
Add to cart
Chemical Identifiers
CAS244193-50-8
IUPAC Name1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular FormulaC10H19BF4N2
InChI KeyMFXLOVLEQJRXFP-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4285-1.4315 @ 20?C
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Assay by NMR≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only