Thermo Scientific Chemicals

N-Allylmethylamine, 96%, Thermo Scientific Chemicals

Catalog number: H27551.03
1 g, Each
Thermo Scientific Chemicals

N-Allylmethylamine, 96%, Thermo Scientific Chemicals

Catalog number: H27551.03
1 g, Each
Quantity
Catalog number: H27551.03
also known as H27551-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
627-37-2
IUPAC Name
methyl(prop-2-en-1-yl)amine
Molecular Formula
C4H9N
InChI Key
IOXXVNYDGIXMIP-UHFFFAOYSA-N
SMILES
CNCC=C
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4095-1.4155 @ 20?C
Form
Liquid
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms

Description

N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text