Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium ethyl sulfate, 99%, Thermo Scientific Chemicals

Catalog number: H27571.30
250 g, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium ethyl sulfate, 99%, Thermo Scientific Chemicals

Catalog number: H27571.30
250 g, Each
Quantity
Catalog number: H27571.30
also known as H27571-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
342573-75-5
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium ethyl sulfate
Molecular Formula
C8H16N2O4S
InChI Key
VRFOKYHDLYBVAL-UHFFFAOYSA-M
SMILES
CCOS([O-])(=O)=O.CCN1C=C[N+](C)=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Ion Chromatography
≥98.5%
Water Content (Karl Fischer Titration)
≤1,500 ppm

Description

1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.

Solubility
Miscible with water and alcohol.

Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text