Thermo Scientific Chemicals

4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27578.03
1 g, Each
Thermo Scientific Chemicals

4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27578.03
1 g, Each
Quantity
Catalog number: H27578.03
also known as H27578-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
172843-97-9
IUPAC Name
4-{[(tert-butoxy)carbonyl]amino}oxane-4-carboxylate
Molecular Formula
C11H18NO5
InChI Key
SPPDKPRJPFTBEV-UHFFFAOYSA-M
SMILES
CC(C)(C)OC(=O)NC1(CCOCC1)C([O-])=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%

Description

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. (15 g/L) (25°C)

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text