2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals
2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H27599.03
also known as H27599-03
Price (USD)/ Each
606.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
606.00
Add to cart
2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals
Catalog numberH27599.03
Price (USD)/ Each
606.00
-
Add to cart
Chemical Identifiers
CAS458532-94-0
IUPAC Name(2-bromopyridin-4-yl)boronic acid
Molecular FormulaC5H5BBrNO2
InChI KeyDFIKFQMGZNTFLI-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Br)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formpowder
Assay (GC, Pinacol ester)> 94.0%
Proton NMRconforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only