2-Cyano-6-fluoropyridine, 95%, Thermo Scientific Chemicals
2-Cyano-6-fluoropyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyano-6-fluoropyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number H27600.03
also known as H27600-03
Price (USD)/ Each
265.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
265.00
Add to cart
2-Cyano-6-fluoropyridine, 95%, Thermo Scientific Chemicals
Catalog numberH27600.03
Price (USD)/ Each
265.00
-
Add to cart
Chemical Identifiers
CAS3939-15-9
IUPAC Name6-fluoropyridine-2-carbonitrile
Molecular FormulaC6H3FN2
InChI KeyNVOLTPVZQXTZCW-UHFFFAOYSA-N
SMILESFC1=CC=CC(=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥94.0%
Melting Point (clear melt)27.0-40.0?C
It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Yinsheng Zhang.; Laura Greenfield. ;Yang Hong. Syntheses of dual-radioisotope-labeled CP-I, a GABAA receptor partial agonist.Journal of Labelled Compounds and Radiopharmaceuticals. 2011, 54 (8),411-417 .
  2. Teruo Umemoto.; Ginjiro Tomizawa. Preparation of 2-fluoropyridines via base-induced decomposition of N-fluoropyridinium salts.J. Org. Chem. 1989, 54 (7),1726-1731 .