Thermo Scientific Chemicals

(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H27612.MD
250 mg, Each
Thermo Scientific Chemicals

(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H27612.MD
250 mg, Each
Quantity
Catalog number: H27612.MD
also known as H27612-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85977-52-2
IUPAC Name
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Formula
C11H12O2
InChI Key
VDLWTJCSPSUGOA-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCC2=CC=CC=C12
Optical Rotation
-11.5? ± 1.0? (c=1 in methanol)
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
53.5-59.5?C
Appearance (Color)
Cream
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text