Thermo Scientific Chemicals

Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H27614.03
1 g, Each
Thermo Scientific Chemicals

Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H27614.03
1 g, Each
Quantity
Catalog number: H27614.03
also known as H27614-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1458-18-0
IUPAC Name
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Molecular Formula
C6H5Cl2N3O2
InChI Key
USYMCUGEGUFUBI-UHFFFAOYSA-N
SMILES
COC(=O)C1=NC(Cl)=C(Cl)N=C1N
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
227-241?C
Appearance (Color)
Red to pale brown to dark brown
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text