Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H27614.06
also known as H27614-06
Price (USD)/ Each
386.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
386.00
Add to cart
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH27614.06
Price (USD)/ Each
386.00
-
Add to cart
Chemical Identifiers
CAS1458-18-0
IUPAC Namemethyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Molecular FormulaC6H5Cl2N3O2
InChI KeyUSYMCUGEGUFUBI-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC(Cl)=C(Cl)N=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to pale brown to dark brown
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)227-241?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only