1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
Venier, O.; Pascal, C.; Braun, A.; Namane, C.; Mougenot, P.; Crespin, O.; Pacquet, F.; Mougenot, C.; Monseau, C.; Onofri, B.; Faïhun, R. D.; Leger, C.; Hassine, M. B. H.; Pham, T. V.; Ragot, J. L.; Philippo, C.; Gussregen, S.; Engel, C.; Farjot, G,; Noah, L.; Maniani, K.; Nicolai, E. Pyrrolidine-pyrazole ureas as potent and selective inhibitors of 11 β-hydroxysteroid-dehydrogenase type 1. Bioorg. Med. Chem. Lett. 2011, 21 (8), 2244-2251.