Thermo Scientific Chemicals

1H-Pyrazole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27619.06
5 g, Each
Thermo Scientific Chemicals

1H-Pyrazole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27619.06
5 g, Each
Quantity
Catalog number: H27619.06
also known as H27619-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
844501-71-9
IUPAC Name
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Formula
C9H15BN2O2
InChI Key
KWLOIDOKWUESNM-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=NN1
Appearance (Color)
White
Form
Powder
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
102.0-116.0?C

Description

1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text