Thermo Scientific Chemicals

Ethyl cellulose

Catalog number: H27621.MD
250 mg, Each
Thermo Scientific Chemicals

Ethyl cellulose

Catalog number: H27621.MD
250 mg, Each
Quantity
Catalog number: H27621.MD
also known as H27621-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-57-3
IUPAC Name
2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular Formula
C23H24N6O4
InChI Key
ARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILES
CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
Form
Powder or granules
Appearance (Color)
White to cream to yellow to pale brown
Assay (unspecified)
44.0-51.0% ethoxy groups
Viscosity
8-12 cP (5% in 80/20 toluene/ethanol, 25?C)

Description

Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color. In addition to this, it is used in lacquers and plastic toughing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text