1-(1-Methyl-4-piperidinyl)piperazine, 98%, Thermo Scientific Chemicals
1-(1-Methyl-4-piperidinyl)piperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(1-Methyl-4-piperidinyl)piperazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27624.03
also known as H27624-03
Price (USD)/ Each
49.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
49.70
Add to cart
1-(1-Methyl-4-piperidinyl)piperazine, 98%, Thermo Scientific Chemicals
Catalog numberH27624.03
Price (USD)/ Each
49.70
-
Add to cart
Chemical Identifiers
CAS23995-88-2
IUPAC Name1-(1-methylpiperidin-4-yl)piperazine
Molecular FormulaC10H21N3
InChI KeyOHUMKYGINIODOY-UHFFFAOYSA-N
SMILESCN1CCC(CC1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormFused solid
Assay (GC)≥97.5%
Melting Point (clear melt)25.0-32.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only