3-Bromo-2-cyanopyridine, 98%, Thermo Scientific Chemicals
3-Bromo-2-cyanopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-2-cyanopyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H27632.03
also known as H27632-03
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
3-Bromo-2-cyanopyridine, 98%, Thermo Scientific Chemicals
Catalog numberH27632.03
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS55758-02-6
IUPAC Name3-bromopyridine-2-carbonitrile
Molecular FormulaC6H3BrN2
InChI KeyHCOPIUVJCIZALB-UHFFFAOYSA-N
SMILESBrC1=CC=CN=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only