Thermo Scientific Chemicals

1,1':3',1″-Terphenyl-5'-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27640.03
1 g, Each
Thermo Scientific Chemicals

1,1':3',1″-Terphenyl-5'-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27640.03
1 g, Each
Quantity
Catalog number: H27640.03
also known as H27640-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128388-54-5
IUPAC Name
{5-phenyl-[1,1'-biphenyl]-3-yl}boronic acid
Molecular Formula
C18H15BO2
InChI Key
MRBZYVMZUBUDAX-UHFFFAOYSA-N
SMILES
OB(O)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥94.0%
Assay (HPLC)
≥94.0%
Proton NMR
Conforms to structure

Description

A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

Solubility
Insoluble in water.

Notes
Refrigerated (2-4°C). Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text