Thermo Scientific Chemicals

2,4-Dichloro-5-nitropyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H27644.09
10 g, Each
Thermo Scientific Chemicals

2,4-Dichloro-5-nitropyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H27644.09
10 g, Each
Quantity
Catalog number: H27644.09
also known as H27644-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
49845-33-2
IUPAC Name
2,4-dichloro-5-nitropyrimidine
Molecular Formula
C4HCl2N3O2
InChI Key
INUSQTPGSHFGHM-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CN=C(Cl)N=C1Cl
Form
Fused solid or liquid as melt
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
24.0-34.0?C
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text