Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: H27651.06
5 g, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: H27651.06
5 g, Each
Quantity
Catalog number: H27651.06
also known as H27651-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65039-09-0
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium chloride
Molecular Formula
C6H11ClN2
InChI Key
BMQZYMYBQZGEEY-UHFFFAOYSA-M
SMILES
[Cl-].CCN1C=C[N+](C)=C1
Appearance (Color)
White to cream to yellow to orange
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
<1.0%

Description

Used in cellulose processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in cellulose processing.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text