Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only
General References:
- Popp, S.; Bösmann, A.; Wölfel, R.; Wasserscheid, P. Screening of Ionic Liquid/H2O Working Pairs for Application in Low Temperature Driven Sorption Heat Pump Systems. ACS Sustainable Chem. Eng. 2015, 3 (4), 750-757.
- Okamura, T. A.; Furuya, R.; Onitsuka, K. Synthesis and structures of soluble magnesium and zinc carboxylates containing intramolecular NH···O hydrogen bonds in nonpolar solvents. Dalton Trans. 2015, 44 (16), 7512-7523.