Thermo Scientific Chemicals

Tetramethylammonium acetate, tech. 90%, Thermo Scientific Chemicals

Catalog number: H27659.14
25 g, Each
Thermo Scientific Chemicals

Tetramethylammonium acetate, tech. 90%, Thermo Scientific Chemicals

Catalog number: H27659.14
25 g, Each
Quantity
Catalog number: H27659.14
also known as H27659-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10581-12-1
IUPAC Name
tetramethylazanium acetate
Molecular Formula
C6H15NO2
InChI Key
MRYQZMHVZZSQRT-UHFFFAOYSA-M
SMILES
CC([O-])=O.C[N+](C)(C)C
Appearance (Color)
White
Form
crystalline powder
Assay (Non-aqueous acid-base Titration)
> 88.0%
Infrared spectrum
conforms to structure
Water Content (Karl Fischer Titration)
< 5.0%

Description

Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text