This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylamine is used as a precursor in the preparation of nitrosodimethylamine.
Solubility
Miscible with water and most organic solvents.
Notes
Hygroscopic. Incompatible with oxidizing agents and oxygen,Sodium aluminum hydride, sodium tetrahydroaluminate, lithium tetrahydroaluminate, borane, metal halides, strong acids, caustic alkalis and bromine.
RUO – Research Use Only
Jochum, F. D.; Theato, P. Temperature and light sensitive copolymers containing azobenzene moieties prepared via a polymer analogous. Polymer 2009, 50 (14), 2730-2735.
Lee, B. K.; Biscoe, M. R.; Buchwald, S. L. Simple, efficient protocols for the Pd-catalyzed cross-coupling reaction of aryl chlorides and dimethylamine. Tetrahedron Lett. 2009, 50 (26), 3672-3674.