It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.
Solubility
Slightly soluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
I. Ernest.; J. Kalvoda.; G. Rihs.; M. Mutterd. Three novel mimics for the construction of sterically constrained protein turn models.Tetrahedron Lett. 1990, 31 (28), 4011-4014 .