4-Amino-5-bromo-2-chloropyridine, 95%, Thermo Scientific Chemicals
4-Amino-5-bromo-2-chloropyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-5-bromo-2-chloropyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H27674.03
also known as H27674-03
Price (USD)/ Each
295.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
295.00
Add to cart
4-Amino-5-bromo-2-chloropyridine, 95%, Thermo Scientific Chemicals
Catalog numberH27674.03
Price (USD)/ Each
295.00
-
Add to cart
Chemical Identifiers
CAS857730-21-3
IUPAC Name5-bromo-2-chloropyridin-4-amine
Molecular FormulaC5H4BrClN2
InChI KeyMGZWZNBANVSZLM-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)122.0-134.0?C
Appearance (Color)White to pale cream
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only