1-Methyl-3-n-propylimidazolium iodide, 98%, Thermo Scientific Chemicals
1-Methyl-3-n-propylimidazolium iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-3-n-propylimidazolium iodide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number H27682.18
also known as H27682-18
Price (USD)/ Each
233.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
233.00
Add to cart
1-Methyl-3-n-propylimidazolium iodide, 98%, Thermo Scientific Chemicals
Catalog numberH27682.18
Price (USD)/ Each
233.00
-
Add to cart
Chemical Identifiers
CAS119171-18-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
Identification (FTIR)Conforms
Assay (Titration ex Iodide)≥97.5 to ≤102.5%
Refractive Index1.5885-1.5930 @ 20?C
FormLiquid or viscous liquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only