Thermo Scientific Chemicals

Methyl 4-methoxyindole-2-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: H27743.MD
250 mg, Each
Thermo Scientific Chemicals

Methyl 4-methoxyindole-2-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: H27743.MD
250 mg, Each
Quantity
Catalog number: H27743.MD
also known as H27743-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111258-23-2
IUPAC Name
methyl 4-methoxy-1H-indole-2-carboxylate
Molecular Formula
C11H11NO3
InChI Key
GLCZQTLCVLVFGV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC2=C(OC)C=CC=C2N1
Form
Powder
Melting Point (clear melt)
138.5-144.5?C
Assay (GC)
≥98.5%
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text