N(alpha)-Boc-D-tryptophanol, 98%
N(alpha)-Boc-D-tryptophanol, 98%
Thermo Scientific Chemicals

N(alpha)-Boc-D-tryptophanol, 98%

Have Questions?
Quantity:
1 g
Catalog number H27763.03
also known as H27763-03
Price (USD)/ Each
268.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
268.00
Add to cart
N(alpha)-Boc-D-tryptophanol, 98%
Catalog numberH27763.03
Price (USD)/ Each
268.00
-
Add to cart
Chemical Identifiers
CAS158932-00-4
IUPAC Nametert-butyl N-[(2R)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Molecular FormulaC16H22N2O3
InChI KeyJEFQUFUAEKORKL-GFCCVEGCSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)117.0-124.0?C
Appearance (Color)Pale cream
Assay (HPLC)≥97.5%
Optical Rotation53.4 ± 0.1? (C=2 in acetic acid)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only