This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Maud Reiter.; Sandrine Ropp.; Véronique Gouverneur. Oxidative Cyclization of β-Hydroxyenones with Palladium(II): A Novel Entry to 2,3-Dihydro-4H-pyran-4-ones.Org. Lett. 2004, 6 (1),91-94 .
- Yoshitomo Hamuro.; Steven J. Geib.; Andrew D. Hamilton. Oligoanthranilamides. Non-Peptide Subunits That Show Formation of Specific Secondary Structure.J. Am. Chem. Soc. 1996, 118 (32),7529-7541 .