Thermo Scientific Chemicals

Bis(tetramethylcyclopentadienyl)nickel(II), 98+%, Thermo Scientific Chemicals

Catalog number: H27784.06
5 g, Each
Thermo Scientific Chemicals

Bis(tetramethylcyclopentadienyl)nickel(II), 98+%, Thermo Scientific Chemicals

Catalog number: H27784.06
5 g, Each
Quantity
Catalog number: H27784.06
also known as H27784-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79019-60-6
IUPAC Name
nickel(2+) bis(2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide)
Molecular Formula
C18H26Ni
InChI Key
PPBIVTWBQQUEKM-UHFFFAOYNA-N
SMILES
[Ni++].CC1[C-]=C(C)C(C)=C1C.CC1[C-]=C(C)C(C)=C1C
Assay (unspecified)
≥98.0 to ≤102.0% base on Carbon Elemental Analysis
Elemental Analysis
Hydrogen H: 8.53-8.88% (Theoretical 8.70%)
Appearance (Color)
Green to black
Elemental Analysis
Carbon C: 70.37-73.24% (Theoretical 71.80%)
Form
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text