Ligand for ruthenium-catalyzed greener amide bond formation by dehydrogenative coupling of amines and alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ligand for ruthenium-catalyzed greener amide bond formation by dehydrogenative coupling of amines and alcohols.
Solubility
Soluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only
General References:
- Ian R. Baxendale Dr.,; Steven V. Ley Prof.,; Andrew C. Mansfield2,; Christopher D. Smith Dr. Multistep Synthesis Using Modular Flow Reactors: Bestmann-Ohira Reagent for the Formation of Alkynes and Triazoles. Angewandte Chemie . 2009, 48(22),4017-4021,.
- .Structural characterization of Group 4 transition metal halide bis-Arduengo carbene complexes MCl4L2:: X-ray crystal structure analyses and DFT calculations. Journal of Organometallic Chemistry. 2002, 663 (1-2), 192-203.