Thermo Scientific Chemicals

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog number: H27827.06
5 g, Each
Thermo Scientific Chemicals

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog number: H27827.06
5 g, Each
Quantity
Catalog number: H27827.06
also known as H27827-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
227617-70-1
Appearance (Color)
White to pale yellow
Assay from Supplier's CofA
≥98.5%
Water Content (Karl Fischer Titration)
<500ppm
Impurity content
Halides: <1000ppm
Melting Point
36-50?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text