1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals
1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number H27827.18
also known as H27827-18
Price (USD)/ Each
305.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
305.00
Add to cart
1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals
Catalog numberH27827.18
Price (USD)/ Each
305.00
-
Add to cart
Chemical Identifiers
CAS227617-70-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay from Supplier's CofA≥98.5%
Water Content (Karl Fischer Titration)<500ppm
Impurity contentHalides: <1000ppm
Melting Point36-50?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only