Thermo Scientific Chemicals

Brivudine, 98%, Thermo Scientific Chemicals

Catalog number: H27853.MF
50 mg, Each
Thermo Scientific Chemicals

Brivudine, 98%, Thermo Scientific Chemicals

Catalog number: H27853.MF
50 mg, Each
Quantity
Catalog number: H27853.MF
also known as H27853-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69304-47-8
IUPAC Name
5-[(1E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C11H13BrN2O5
InChI Key
ODZBBRURCPAEIQ-PIXDULNESA-N
SMILES
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C=CBr)C(=O)NC1=O
Form
Crystalline powder or powder
Proton NMR
Conforms to structure
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥97.5%

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text