Thermo Scientific Chemicals

Dichloro(dimethylamino)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: H27856.06
5 g, Each
Thermo Scientific Chemicals

Dichloro(dimethylamino)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: H27856.06
5 g, Each
Quantity
Catalog number: H27856.06
also known as H27856-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
683-85-2
IUPAC Name
(dichlorophosphanyl)dimethylamine
Molecular Formula
C2H6Cl2NP
InChI Key
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
SMILES
CN(C)P(Cl)Cl
Appearance (Color)
Colorless to pale yellow
Comment
Product may contain a small amount of sediment up on standing which does not affect the overall quality of the product.
Assay by NMR
(by Phosphorous NMR) : >97.5%
Phosphorus-31 NMR
Conforms to structure
Form
Liquid

Description

Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text