Thermo Scientific Chemicals

2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: H27859.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: H27859.06
5 g, Each
Quantity
Catalog number: H27859.06
also known as H27859-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66916-99-2
IUPAC Name
2-(2-bromo-4-methoxyphenyl)acetic acid
Molecular Formula
C9H9BrO3
InChI Key
XQELSBAAFMYSMG-UHFFFAOYSA-N
SMILES
COC1=CC(Br)=C(CC(O)=O)C=C1
Form
Powder
Assay (Silylated GC)
≥96.0%
Melting Point
125.0-134.0?C
Appearance (Color)
White
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text