2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals
2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H27859.14
also known as H27859-14
Price (USD)/ Each
319.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
319.00
Add to cart
2-Bromo-4-methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals
Catalog numberH27859.14
Price (USD)/ Each
319.00
-
Add to cart
Chemical Identifiers
CAS66916-99-2
IUPAC Name2-(2-bromo-4-methoxyphenyl)acetic acid
Molecular FormulaC9H9BrO3
InChI KeyXQELSBAAFMYSMG-UHFFFAOYSA-N
SMILESCOC1=CC(Br)=C(CC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
FormPowder
Assay (Silylated GC)≥96.0%
Melting Point125.0-134.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only