Thermo Scientific Chemicals

2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific Chemicals

Catalog number: H27877.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific Chemicals

Catalog number: H27877.06
5 g, Each
Quantity
Catalog number: H27877.06
also known as H27877-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
95-24-9
IUPAC Name
6-chloro-1,3-benzothiazol-2-amine
Molecular Formula
C7H5ClN2S
InChI Key
VMNXKIDUTPOHPO-UHFFFAOYSA-N
SMILES
NC1=NC2=CC=C(Cl)C=C2S1
Assay (GC)
≥98.5%
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
196.5-200.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell