Methyl thiocyanate, 99%, Thermo Scientific Chemicals
Methyl thiocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl thiocyanate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Catalog number H27879.AC
also known as H27879-AC
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
64.70 
Save 10.05 (16%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
64.70 
Save 10.05 (16%)
Add to cart
Methyl thiocyanate, 99%, Thermo Scientific Chemicals
Catalog numberH27879.AC
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
64.70 
Save 10.05 (16%)
-
Add to cart
Chemical Identifiers
CAS556-64-9
IUPAC Name(methylsulfanyl)carbonitrile
Molecular FormulaC2H3NS
InChI KeyVYHVQEYOFIYNJP-UHFFFAOYSA-N
SMILESCSC#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.46701-1.4700 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only