Thermo Scientific Chemicals

Methyl (+)-4-Boc-piperazine-2-carboxylate, Thermo Scientific Chemicals

Catalog number: H27886.06
5 g, Each
Thermo Scientific Chemicals

Methyl (+)-4-Boc-piperazine-2-carboxylate, Thermo Scientific Chemicals

Catalog number: H27886.06
5 g, Each
Quantity
Catalog number: H27886.06
also known as H27886-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
129799-08-2
Form
powder or fused solid
Water Content (Karl Fischer Titration)
< 2.0%
Appearance (Color)
White to pale yellow or cream
Assay (GC)
> 96.0%

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text